Product Details

SNP ID
rs118204034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89776598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGCGCGCACCGGACCAGCGGGC[G/T]GACAGCGGCAGAGCGCCAGCGAGCG
Phenotype
MIM: 151530 MIM: 605195
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANPEP PubMed Links

Gene Details

Gene
ANPEP
Gene Name
alanyl aminopeptidase, membrane
There are no transcripts associated with this gene.

Gene
MESP2
Gene Name
mesoderm posterior bHLH transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039958.1 241 Missense Mutation GGA,TGA G81* NP_001035047.1

View Full Product Details