Product Details

SNP ID
rs138128066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34361443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACCAGCCACCGTCTGAGGATCA[C/G]AGAGCTGTAGCTGCCTGGCAAACTC
Phenotype
MIM: 612039 MIM: 608963
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LPCAT4 PubMed Links

Gene Details

Gene
LPCAT4
Gene Name
lysophosphatidylcholine acyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153613.2 1196 Missense Mutation TCT,TGT S367C NP_705841.2
XM_006720454.2 1196 Missense Mutation TCT,TGT S367C XP_006720517.1
XM_011521422.1 1196 Missense Mutation TCT,TGT S199C XP_011519724.1
Gene
NUTM1
Gene Name
NUT midline carcinoma family member 1
There are no transcripts associated with this gene.

View Full Product Details