Product Details

SNP ID
rs138215044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89472828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGAGGGCTTGAAGGTGGGGTC[A/C]TCAGGGATGTAGACAGTGCTGTTCC
Phenotype
MIM: 605381
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371031 PubMed Links

Gene Details

Gene
LOC105371031
Gene Name
translation initiation factor IF-2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017022800.1 1416 Intron XP_016878289.1
Gene
RHCG
Gene Name
Rh family C glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321041.1 1416 Missense Mutation GAG,GAT E449D NP_001307970.1
NM_016321.2 1416 Missense Mutation GAG,GAT E449D NP_057405.1

View Full Product Details