Product Details

SNP ID
rs138236822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41517627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCAGAGGGGGCAACTGAGAATAG[A/T]GCTCAAAGCCCTCTGGGAGCGTGGA
Phenotype
MIM: 151520 MIM: 611475
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LTK PubMed Links

Gene Details

Gene
LTK
Gene Name
leukocyte receptor tyrosine kinase
There are no transcripts associated with this gene.

Gene
RPAP1
Gene Name
RNA polymerase II associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015540.3 4511 Missense Mutation CAC,CTC H1366L NP_056355.2
XM_005254297.1 4511 Missense Mutation CAC,CTC H1366L XP_005254354.1
XM_006720469.1 4511 Missense Mutation CAC,CTC H1306L XP_006720532.1

View Full Product Details