Product Details

SNP ID
rs138263972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65898776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTCGACAAGGATGGCACATCTG[C/T]GTACGGAGACCCCATGTGCACAGGC
Phenotype
MIM: 612454 MIM: 605570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEGF11 PubMed Links

Gene Details

Gene
MEGF11
Gene Name
multiple EGF like domains 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032445.2 3496 Missense Mutation ACA,GCA T976A NP_115821.2
XM_017022670.1 3496 Missense Mutation ACA,GCA T1029A XP_016878159.1
XM_017022671.1 3496 Missense Mutation ACA,GCA T1072A XP_016878160.1
XM_017022672.1 3496 Missense Mutation ACA,GCA T976A XP_016878161.1
XM_017022673.1 3496 Intron XP_016878162.1
XM_017022674.1 3496 Missense Mutation ACA,GCA T786A XP_016878163.1
XM_017022675.1 3496 Missense Mutation ACA,GCA T786A XP_016878164.1
Gene
RAB11A
Gene Name
RAB11A, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details