Product Details

SNP ID
rs138345086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73998443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAAGACCAACAACATCTTCTGCT[C/T]CAACCCCAACCACCGCACCCCTACG
Phenotype
MIM: 102578 MIM: 608326
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PML PubMed Links

Gene Details

Gene
PML
Gene Name
promyelocytic leukemia
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002675.3 709 Missense Mutation TCC,TTC S190F NP_002666.1
NM_033238.2 709 Missense Mutation TCC,TTC S190F NP_150241.2
NM_033239.2 709 Missense Mutation TCC,TTC S190F NP_150242.1
NM_033240.2 709 Missense Mutation TCC,TTC S190F NP_150243.2
NM_033244.3 709 Missense Mutation TCC,TTC S190F NP_150247.2
NM_033246.2 709 Missense Mutation TCC,TTC S190F NP_150249.1
NM_033247.2 709 Missense Mutation TCC,TTC S190F NP_150250.2
NM_033249.2 709 Missense Mutation TCC,TTC S190F NP_150252.1
NM_033250.2 709 Missense Mutation TCC,TTC S190F NP_150253.2
Gene
STOML1
Gene Name
stomatin like 1
There are no transcripts associated with this gene.

View Full Product Details