Product Details

SNP ID
rs138507615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84643714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCATCATAGACCTGGACTGTA[A/C]CATCAAAACCTGAGAATACAGAAAA
Phenotype
MIM: 610417 MIM: 616144
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCAND2P PubMed Links

Gene Details

Gene
SCAND2P
Gene Name
SCAN domain containing 2 pseudogene
There are no transcripts associated with this gene.

Gene
WDR73
Gene Name
WD repeat domain 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032856.3 962 Missense Mutation NP_116245.2

View Full Product Details