Product Details

SNP ID
rs138592907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42946486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCTGCTTTGTACCCTTGTGGAA[C/T]AACTAAGATTCTCAGTCATTAACTA
Phenotype
MIM: 605981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBR1 PubMed Links

Gene Details

Gene
UBR1
Gene Name
ubiquitin protein ligase E3 component n-recognin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174916.2 Intron NP_777576.1

View Full Product Details