Product Details

SNP ID
rs138827129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50182816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGCTGGTGAAGCTGGGCTGTG[C/T]CATGGCGTGCCTCAATTACAACATC
Phenotype
MIM: 609123 MIM: 603247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 621 Intron NP_079113.2
XM_011522046.2 621 Intron XP_011520348.1
XM_011522047.2 621 Intron XP_011520349.1
XM_011522048.1 621 Intron XP_011520350.1
XM_011522049.2 621 Intron XP_011520351.1
XM_011522051.2 621 Intron XP_011520353.1
XM_011522052.2 621 Intron XP_011520354.1
XM_011522053.1 621 Intron XP_011520355.1
XM_011522056.2 621 Intron XP_011520358.2
XM_011522058.2 621 Intron XP_011520360.1
XM_011522059.1 621 Intron XP_011520361.1
XM_011522060.1 621 Intron XP_011520362.1
XM_011522061.1 621 Intron XP_011520363.1
XM_011522062.1 621 Intron XP_011520364.1
XM_011522063.1 621 Intron XP_011520365.1
XM_011522064.1 621 Intron XP_011520366.1
XM_011522069.2 621 Intron XP_011520371.1
XM_011522070.1 621 Intron XP_011520372.1
XM_017022587.1 621 Intron XP_016878076.1
XM_017022588.1 621 Intron XP_016878077.1
XM_017022589.1 621 Intron XP_016878078.1
XM_017022590.1 621 Intron XP_016878079.1
XM_017022591.1 621 Intron XP_016878080.1
XM_017022592.1 621 Intron XP_016878081.1
XM_017022593.1 621 Intron XP_016878082.1
XM_017022594.1 621 Intron XP_016878083.1
XM_017022595.1 621 Intron XP_016878084.1
XM_017022596.1 621 Intron XP_016878085.1
XM_017022597.1 621 Intron XP_016878086.1
XM_017022598.1 621 Intron XP_016878087.1
XM_017022599.1 621 Intron XP_016878088.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159629.1 621 Missense Mutation GCC,GTC A130V NP_001153101.1
NM_003645.3 621 Missense Mutation GCC,GTC A130V NP_003636.2

View Full Product Details