Product Details

SNP ID
rs138870728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84816894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGCTGGGCCAACAGCCACTGGC[A/G]AGGCAAGGCGAGGGTCAGTCACGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALPK3 PubMed Links

Gene Details

Gene
ALPK3
Gene Name
alpha kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020778.4 215 Silent Mutation GCA,GCG A16A NP_065829.3
XM_017022444.1 215 Silent Mutation GCA,GCG A16A XP_016877933.1
XM_017022445.1 215 Silent Mutation GCA,GCG A16A XP_016877934.1

View Full Product Details