Product Details

SNP ID
rs139012996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:69416058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCAAACGAACCTTAAAGACCCA[A/G]TTGGGGTAAGGTATCCCTGTAAATT
Phenotype
MIM: 605064 MIM: 607781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF23 PubMed Links

Gene Details

Gene
KIF23
Gene Name
kinesin family member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281301.1 245 UTR 5 NP_001268230.1
NM_004856.6 245 Missense Mutation ATT,GTT I26V NP_004847.2
NM_138555.3 245 Missense Mutation ATT,GTT I26V NP_612565.1
XM_005254796.2 245 Missense Mutation ATT,GTT I26V XP_005254853.1
XM_005254797.2 245 Missense Mutation ATT,GTT I26V XP_005254854.1
XM_005254798.2 245 Missense Mutation ATT,GTT I26V XP_005254855.1
XM_005254799.2 245 Missense Mutation ATT,GTT I26V XP_005254856.1
XM_011522238.1 245 UTR 5 XP_011520540.1
XM_011522239.2 245 UTR 5 XP_011520541.1
XM_011522240.1 245 Intron XP_011520542.1
XM_017022733.1 245 Missense Mutation ATT,GTT I26V XP_016878222.1
Gene
LOC145694
Gene Name
uncharacterized LOC145694
There are no transcripts associated with this gene.

Gene
PAQR5
Gene Name
progestin and adipoQ receptor family member 5
There are no transcripts associated with this gene.

View Full Product Details