Product Details

SNP ID
rs139285421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89582827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCTGCTCAGGAGTGGAATAAAG[C/T]TGTCAAGTGAACCTCATCTTTCTCC
Phenotype
MIM: 613298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TICRR PubMed Links

Gene Details

Gene
TICRR
Gene Name
TOPBP1 interacting checkpoint and replication regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308025.1 796 Silent Mutation CTG,TTG L266L NP_001294954.1
NM_152259.3 796 Silent Mutation CTG,TTG L266L NP_689472.3

View Full Product Details