Product Details

SNP ID
rs139335305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:81102891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTGGAAGGGAAGAATTTGATCT[A/G]TGGGGTTGAATCAATGAGGAAGTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP161 PubMed Links

Gene Details

Gene
CFAP161
Gene Name
cilia and flagella associated protein 161
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173528.2 Intron NP_775799.2
XM_006720408.2 Intron XP_006720471.1
XM_011521280.1 Intron XP_011519582.1
XM_017021963.1 Intron XP_016877452.1

View Full Product Details