Product Details

SNP ID
rs139663241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:82042488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTGCAATATGAAATATAATTTT[A/T]CAAGTCAAAAAAGAATAAAATACTT
Phenotype
MIM: 611008
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MEX3B PubMed Links

Gene Details

Gene
MEX3B
Gene Name
mex-3 RNA binding family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032246.4 2820 UTR 3 NP_115622.2

View Full Product Details