Product Details

SNP ID
rs139694386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41828174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCGGAGCGAGTTACGAGAATTCC[C/T]GGCCGCTGCAAGGGGTGAGTGGCTC
Phenotype
MIM: 616786
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JMJD7 PubMed Links

Gene Details

Gene
JMJD7
Gene Name
jumonji domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114632.1 90 Missense Mutation CCG,CTG P17L NP_001108104.1
Gene
JMJD7-PLA2G4B
Gene Name
JMJD7-PLA2G4B readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198588.1 90 Missense Mutation CCG,CTG P17L NP_001185517.1
NM_005090.3 90 Missense Mutation CCG,CTG P17L NP_005081.1
Gene
MAPKBP1
Gene Name
mitogen-activated protein kinase binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128608.1 90 Intron NP_001122080.1
NM_001265611.1 90 Intron NP_001252540.1
NM_014994.2 90 Intron NP_055809.2
XM_006720438.1 90 Intron XP_006720501.1
XM_006720439.2 90 Intron XP_006720502.1
XM_011521383.1 90 Intron XP_011519685.1
XM_011521384.2 90 Intron XP_011519686.1
XM_017022017.1 90 Intron XP_016877506.1

View Full Product Details