Product Details

SNP ID
rs139874813
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79898996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGAAAAATTAGGCCGGTTTCAC[A/G]ATATGGAGTGTCCTTTCTGGTCAAC
Phenotype
MIM: 604197
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MTHFS PubMed Links
Additional Information
For this assay, SNP(s) [rs7257] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MTHFS
Gene Name
5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase)
There are no transcripts associated with this gene.

Gene
ST20
Gene Name
suppressor of tumorigenicity 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100879.1 855 Missense Mutation CGT,TGT R59C NP_001094349.1
NM_001100880.2 855 Missense Mutation CGT,TGT R59C NP_001094350.1
NM_001199757.1 855 Missense Mutation CGT,TGT R59C NP_001186686.1
Gene
ST20-MTHFS
Gene Name
ST20-MTHFS readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199760.1 855 Intron NP_001186689.1

View Full Product Details