Product Details

SNP ID
rs139875923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64154429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGAATGGTGTGCTCCAGGGCC[C/T]CTACAGCTTCAGCATCAGCCCAGAT
Phenotype
MIM: 123841
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPIB PubMed Links

Gene Details

Gene
PPIB
Gene Name
peptidylprolyl isomerase B
There are no transcripts associated with this gene.

Gene
SNX22
Gene Name
sorting nexin 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024798.2 562 Missense Mutation CCC,CTC P168L NP_079074.2
XM_005254677.3 562 Intron XP_005254734.1
XM_017022581.1 562 Missense Mutation CCC,CTC P140L XP_016878070.1

View Full Product Details