Product Details
- SNP ID
-
rs140351413
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:58880046 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTTAAATGGCTTAAATCCGGAGC[C/T]ACTTCCTCTTGGCATGGAATTGCCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLTM
PubMed Links
Gene Details
- Gene
- SLTM
- Gene Name
- SAFB like transcription modulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001013843.2 |
2965 |
Missense Mutation |
AGC,GGC |
S1002G |
NP_001013865.1 |
NM_024755.3 |
2965 |
Missense Mutation |
AGC,GGC |
S1020G |
NP_079031.2 |
XM_006720686.3 |
2965 |
Missense Mutation |
AGC,GGC |
S1073G |
XP_006720749.3 |
XM_006720690.1 |
2965 |
Missense Mutation |
AGC,GGC |
S589G |
XP_006720753.1 |
XM_011522022.1 |
2965 |
Missense Mutation |
AGC,GGC |
S1162G |
XP_011520324.1 |
XM_011522023.1 |
2965 |
Missense Mutation |
AGC,GGC |
S1144G |
XP_011520325.1 |
XM_011522024.1 |
2965 |
Missense Mutation |
AGC,GGC |
S1112G |
XP_011520326.1 |
XM_011522025.1 |
2965 |
Missense Mutation |
AGC,GGC |
S1096G |
XP_011520327.1 |
XM_011522026.2 |
2965 |
Missense Mutation |
AGC,GGC |
S1089G |
XP_011520328.2 |
XM_011522027.1 |
2965 |
Missense Mutation |
AGC,GGC |
S1080G |
XP_011520329.1 |
XM_011522028.1 |
2965 |
Missense Mutation |
AGC,GGC |
S1062G |
XP_011520330.1 |
XM_011522029.2 |
2965 |
Missense Mutation |
AGC,GGC |
S974G |
XP_011520331.1 |
XM_011522030.2 |
2965 |
Intron |
|
|
XP_011520332.1 |
XM_011522031.2 |
2965 |
Missense Mutation |
AGC,GGC |
S792G |
XP_011520333.1 |
XM_017022576.1 |
2965 |
Missense Mutation |
AGC,GGC |
S952G |
XP_016878065.1 |
XM_017022577.1 |
2965 |
Missense Mutation |
AGC,GGC |
S1071G |
XP_016878066.1 |
XM_017022578.1 |
2965 |
Missense Mutation |
AGC,GGC |
S1055G |
XP_016878067.1 |
XM_017022579.1 |
2965 |
Missense Mutation |
AGC,GGC |
S967G |
XP_016878068.1 |
XM_017022580.1 |
2965 |
Missense Mutation |
AGC,GGC |
S589G |
XP_016878069.1 |
View Full Product Details