Product Details

SNP ID
rs140599842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41502048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCGTGAGCGGCCCAAGCTGCG[A/G]AAGGACATGTACAAGAAAATGCTGG
Phenotype
MIM: 147521 MIM: 151520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITPKA PubMed Links

Gene Details

Gene
ITPKA
Gene Name
inositol-trisphosphate 3-kinase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002220.2 925 Silent Mutation CGA,CGG R285R NP_002211.1
XM_011521522.2 925 Silent Mutation CGA,CGG R113R XP_011519824.1
Gene
LTK
Gene Name
leukocyte receptor tyrosine kinase
There are no transcripts associated with this gene.

View Full Product Details