Product Details

SNP ID
rs140607190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:98969546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCAGGTAGGCACACGCCGCCCT[C/T]GGGCCAGAAGCTGCGGATGTCGGCG
Phenotype
MIM: 147370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGF1R PubMed Links

Gene Details

Gene
IGF1R
Gene Name
insulin like growth factor 1 receptor
There are no transcripts associated with this gene.

Gene
PGPEP1L
Gene Name
pyroglutamyl-peptidase I-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102612.2 551 Missense Mutation AAG,GAG K84E NP_001096082.2
NM_001167902.1 551 Intron NP_001161374.1
XM_011521254.1 551 Missense Mutation AAG,GAG K175E XP_011519556.1
XM_017021933.1 551 Missense Mutation AAG,GAG K30E XP_016877422.1

View Full Product Details