Product Details

SNP ID
rs140908103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79311324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCGCCGGGCCGAGCAGCCCTG[C/T]GGGGCCGAGCTCACCTTCGAGCTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMED3 PubMed Links

Gene Details

Gene
TMED3
Gene Name
transmembrane p24 trafficking protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301203.1 263 Silent Mutation TGC,TGT C25C NP_001288132.1
NM_007364.3 263 Silent Mutation TGC,TGT C25C NP_031390.1
XM_017022040.1 263 Silent Mutation TGC,TGT C25C XP_016877529.1
XM_017022041.1 263 Silent Mutation TGC,TGT C25C XP_016877530.1

View Full Product Details