Product Details

SNP ID
rs141170560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40737971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTGAAACGCAGCTTACCTGATA[A/G]CACCACCTGCCAAGAAGAAAGTGAG
Phenotype
MIM: 179617 MIM: 611873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD51 PubMed Links

Gene Details

Gene
RAD51
Gene Name
RAD51 recombinase
There are no transcripts associated with this gene.

Gene
RMDN3
Gene Name
regulator of microtubule dynamics 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304802.1 1481 Silent Mutation TGC,TGT C373C NP_001291731.1
NM_001323894.1 1481 Silent Mutation TGC,TGT C373C NP_001310823.1
NM_001323895.1 1481 Silent Mutation TGC,TGT C244C NP_001310824.1
NM_001323896.1 1481 Silent Mutation TGC,TGT C399C NP_001310825.1
NM_001323897.1 1481 Silent Mutation TGC,TGT C399C NP_001310826.1
NM_018145.2 1481 Silent Mutation TGC,TGT C373C NP_060615.1
XM_011521755.2 1481 Silent Mutation TGC,TGT C167C XP_011520057.1

View Full Product Details