Product Details
- SNP ID
-
rs141340447
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:96784164 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCTCTTCTGCAGAACCTCGTCGC[A/G]ACATTTCTCAGAAGCCATGACATGT
- Phenotype
-
MIM: 613948
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPATA8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3812907] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPATA8
- Gene Name
- spermatogenesis associated 8
- Gene
- SPATA8-AS1
- Gene Name
- SPATA8 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details