Product Details

SNP ID
rs141544943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73998232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCAGCGGCGCCTGTCGGTGTAC[C/T]GGCAGATTGTGGATGCGCAGGCTGT
Phenotype
MIM: 102578 MIM: 608326
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PML PubMed Links

Gene Details

Gene
PML
Gene Name
promyelocytic leukemia
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002675.3 498 Missense Mutation CGG,TGG R120W NP_002666.1
NM_033238.2 498 Missense Mutation CGG,TGG R120W NP_150241.2
NM_033239.2 498 Missense Mutation CGG,TGG R120W NP_150242.1
NM_033240.2 498 Missense Mutation CGG,TGG R120W NP_150243.2
NM_033244.3 498 Missense Mutation CGG,TGG R120W NP_150247.2
NM_033246.2 498 Missense Mutation CGG,TGG R120W NP_150249.1
NM_033247.2 498 Missense Mutation CGG,TGG R120W NP_150250.2
NM_033249.2 498 Missense Mutation CGG,TGG R120W NP_150252.1
NM_033250.2 498 Missense Mutation CGG,TGG R120W NP_150253.2
Gene
STOML1
Gene Name
stomatin like 1
There are no transcripts associated with this gene.

View Full Product Details