Product Details

SNP ID
rs141946591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50182604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGCGCACCATCCTGCGGGCGTT[A/C]CTGGAGAAAGCGCGCCAGACGCCAC
Phenotype
MIM: 609123 MIM: 603247
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 409 Intron NP_079113.2
XM_011522046.2 409 Intron XP_011520348.1
XM_011522047.2 409 Intron XP_011520349.1
XM_011522048.1 409 Intron XP_011520350.1
XM_011522049.2 409 Intron XP_011520351.1
XM_011522051.2 409 Intron XP_011520353.1
XM_011522052.2 409 Intron XP_011520354.1
XM_011522053.1 409 Intron XP_011520355.1
XM_011522056.2 409 Intron XP_011520358.2
XM_011522058.2 409 Intron XP_011520360.1
XM_011522059.1 409 Intron XP_011520361.1
XM_011522060.1 409 Intron XP_011520362.1
XM_011522061.1 409 Intron XP_011520363.1
XM_011522062.1 409 Intron XP_011520364.1
XM_011522063.1 409 Intron XP_011520365.1
XM_011522064.1 409 Intron XP_011520366.1
XM_011522069.2 409 Intron XP_011520371.1
XM_011522070.1 409 Intron XP_011520372.1
XM_017022587.1 409 Intron XP_016878076.1
XM_017022588.1 409 Intron XP_016878077.1
XM_017022589.1 409 Intron XP_016878078.1
XM_017022590.1 409 Intron XP_016878079.1
XM_017022591.1 409 Intron XP_016878080.1
XM_017022592.1 409 Intron XP_016878081.1
XM_017022593.1 409 Intron XP_016878082.1
XM_017022594.1 409 Intron XP_016878083.1
XM_017022595.1 409 Intron XP_016878084.1
XM_017022596.1 409 Intron XP_016878085.1
XM_017022597.1 409 Intron XP_016878086.1
XM_017022598.1 409 Intron XP_016878087.1
XM_017022599.1 409 Intron XP_016878088.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159629.1 409 Missense Mutation TTA,TTC L59F NP_001153101.1
NM_003645.3 409 Missense Mutation TTA,TTC L59F NP_003636.2

View Full Product Details