Product Details

SNP ID
rs142210120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:23686450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCCCCAAAAGAACTCGTATTC[A/G]GGCGGCTCCACGTATGGGACGCGCT
Phenotype
MIM: 602117
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDN PubMed Links

Gene Details

Gene
NDN
Gene Name
necdin, MAGE family member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002487.2 854 Silent Mutation NP_002478.1

View Full Product Details