Product Details

SNP ID
rs142253131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:57604284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTCTAGGCAAATGTTTGCAGA[A/C]TACGGCTGACCGTACCTCCTGAGAG
Phenotype
MIM: 614071
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GCOM1 PubMed Links

Gene Details

Gene
GCOM1
Gene Name
GRINL1A complex locus 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018090.6 222 Silent Mutation ATA,CTA I31L NP_001018100.1
NM_001018091.6 222 Silent Mutation ATA,CTA I31L NP_001018101.1
NM_001285900.3 222 Silent Mutation ATA,CTA I31L NP_001272829.1
Gene
MYZAP
Gene Name
myocardial zonula adherens protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018100.4 222 Silent Mutation ATA,CTA I31L NP_001018110.1
NM_152451.7 222 Silent Mutation ATA,CTA I31L NP_689664.3

View Full Product Details