Product Details

SNP ID
rs142387462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88639495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTGTGCTGTACGACAAGTTCAT[C/T]CGGCCTGAGGGAGAGATCACCGATT
Phenotype
MIM: 610177 MIM: 604533
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AEN PubMed Links

Gene Details

Gene
AEN
Gene Name
apoptosis enhancing nuclease
There are no transcripts associated with this gene.

Gene
ISG20
Gene Name
interferon stimulated exonuclease gene 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303233.1 343 Silent Mutation ATC,ATT I43I NP_001290162.1
NM_001303234.1 343 Silent Mutation ATC,ATT I43I NP_001290163.1
NM_001303235.1 343 Intron NP_001290164.1
NM_001303236.1 343 UTR 5 NP_001290165.1
NM_001303237.1 343 Silent Mutation ATC,ATT I43I NP_001290166.1
NM_002201.5 343 Silent Mutation ATC,ATT I43I NP_002192.2
XM_005254899.2 343 Silent Mutation ATC,ATT I43I XP_005254956.1
XM_006720488.3 343 Silent Mutation ATC,ATT I43I XP_006720551.1
XM_011521521.2 343 Intron XP_011519823.1
XM_017022147.1 343 UTR 5 XP_016877636.1
XM_017022148.1 343 Silent Mutation ATC,ATT I43I XP_016877637.1

View Full Product Details