Product Details

SNP ID
rs142422927
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63127399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATTTCCCATTTAAGTGGAATTC[A/G]TCATTATGAAAAGGACATAAAAAAG
Phenotype
MIM: 608440
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LACTB PubMed Links

Gene Details

Gene
LACTB
Gene Name
lactamase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288585.1 734 Missense Mutation CAT,CGT H221R NP_001275514.1
NM_032857.4 734 Missense Mutation CAT,CGT H221R NP_116246.2
NM_171846.3 734 Missense Mutation CAT,CGT H221R NP_741982.1
XM_011521180.1 734 Missense Mutation CAT,CGT H221R XP_011519482.1

View Full Product Details