Product Details

SNP ID
rs142598777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89628667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCAAGTCCCGCGTCTCCTCCC[A/G]GGTGCGGGGGGCCCCCTCAGTGAGG
Phenotype
MIM: 611254 MIM: 613298
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF7 PubMed Links

Gene Details

Gene
KIF7
Gene Name
kinesin family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198525.2 3861 Missense Mutation CGG,TGG R1262W NP_940927.2
XM_011521531.2 3861 Missense Mutation CGG,TGG R1303W XP_011519833.1
Gene
TICRR
Gene Name
TOPBP1 interacting checkpoint and replication regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308025.1 3861 Intron NP_001294954.1
NM_152259.3 3861 Intron NP_689472.3

View Full Product Details