Product Details

SNP ID
rs142667711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79970825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTCGTAGAAGTTTCTTGATGA[C/G]AATACCTTCAAATGCAAATATGGTT
Phenotype
MIM: 601056
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BCL2A1 PubMed Links

Gene Details

Gene
BCL2A1
Gene Name
BCL2 related protein A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114735.1 477 Missense Mutation CTC,GTC L99V NP_001108207.1
NM_004049.3 477 Missense Mutation CTC,GTC L99V NP_004040.1

View Full Product Details