Product Details

SNP ID
rs143849049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:82043191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGATGCGGATGGCCTGAGTGACC[A/G]CGGTGTGGCAGACCGGGCACTCGGG
Phenotype
MIM: 611008
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEX3B PubMed Links

Gene Details

Gene
MEX3B
Gene Name
mex-3 RNA binding family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032246.4 2117 Missense Mutation GCG,GTG A560V NP_115622.2

View Full Product Details