Product Details

SNP ID
rs143866600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50182866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGCGAAGTCCCTGCTGCACTGC[C/T]TCCAGTGCTGCGGGGCGAAGGTGCT
Phenotype
MIM: 609123 MIM: 603247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 671 Intron NP_079113.2
XM_011522046.2 671 Intron XP_011520348.1
XM_011522047.2 671 Intron XP_011520349.1
XM_011522048.1 671 Intron XP_011520350.1
XM_011522049.2 671 Intron XP_011520351.1
XM_011522051.2 671 Intron XP_011520353.1
XM_011522052.2 671 Intron XP_011520354.1
XM_011522053.1 671 Intron XP_011520355.1
XM_011522056.2 671 Intron XP_011520358.2
XM_011522058.2 671 Intron XP_011520360.1
XM_011522059.1 671 Intron XP_011520361.1
XM_011522060.1 671 Intron XP_011520362.1
XM_011522061.1 671 Intron XP_011520363.1
XM_011522062.1 671 Intron XP_011520364.1
XM_011522063.1 671 Intron XP_011520365.1
XM_011522064.1 671 Intron XP_011520366.1
XM_011522069.2 671 Intron XP_011520371.1
XM_011522070.1 671 Intron XP_011520372.1
XM_017022587.1 671 Intron XP_016878076.1
XM_017022588.1 671 Intron XP_016878077.1
XM_017022589.1 671 Intron XP_016878078.1
XM_017022590.1 671 Intron XP_016878079.1
XM_017022591.1 671 Intron XP_016878080.1
XM_017022592.1 671 Intron XP_016878081.1
XM_017022593.1 671 Intron XP_016878082.1
XM_017022594.1 671 Intron XP_016878083.1
XM_017022595.1 671 Intron XP_016878084.1
XM_017022596.1 671 Intron XP_016878085.1
XM_017022597.1 671 Intron XP_016878086.1
XM_017022598.1 671 Intron XP_016878087.1
XM_017022599.1 671 Intron XP_016878088.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159629.1 671 Missense Mutation CTC,TTC L147F NP_001153101.1
NM_003645.3 671 Missense Mutation CTC,TTC L147F NP_003636.2

View Full Product Details