Product Details

SNP ID
rs143875418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42069948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGAAGGAGGCATTGACCAGCG[A/G]GAAGTGCAGCAGGATCGGGGCCTCG
Phenotype
MIM: 612864
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G4D PubMed Links

Gene Details

Gene
PLA2G4D
Gene Name
phospholipase A2 group IVD
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178034.3 2286 Missense Mutation CCG,TCG P731S NP_828848.3
XM_011521467.2 2286 Missense Mutation CCG,TCG P675S XP_011519769.1

View Full Product Details