Product Details

SNP ID
rs144189577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72489743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTTCCACAGTTGAGAATCTTAG[C/T]GTATTGCTGTTACCTCAAATTTCCA
Phenotype
MIM: 605624
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARIH1 PubMed Links

Gene Details

Gene
ARIH1
Gene Name
ariadne RBR E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005744.3 Intron NP_005735.2
XM_011521438.2 Intron XP_011519740.1
XM_011521439.2 Intron XP_011519741.1

View Full Product Details