Product Details

SNP ID
rs144247178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88467737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTCATGCAGGCTGTGAGAAACG[C/T]GGGGTCGCGGTTCCTGCGGTCCTGG
Phenotype
MIM: 611851 MIM: 611977
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL46 PubMed Links

Gene Details

Gene
MRPL46
Gene Name
mitochondrial ribosomal protein L46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022163.3 285 Intron NP_071446.2
Gene
MRPS11
Gene Name
mitochondrial ribosomal protein S11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321970.1 285 Missense Mutation GCG,GTG A7V NP_001308899.1
NM_001321972.1 285 UTR 5 NP_001308901.1
NM_001321973.1 285 Missense Mutation GCG,GTG A7V NP_001308902.1
NM_001321974.1 285 Missense Mutation GCG,GTG A7V NP_001308903.1
NM_001321976.1 285 UTR 5 NP_001308905.1
NM_022839.4 285 Missense Mutation GCG,GTG A7V NP_073750.2
NM_176805.3 285 Missense Mutation GCG,GTG A7V NP_789775.1

View Full Product Details