Product Details

SNP ID
rs144279154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:71111256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAAGTGTGGCCTGAAGACTCCC[C/T]GACACTTCATGGGAATCCTTGACAT
Phenotype
MIM: 614476
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CT62 PubMed Links

Gene Details

Gene
CT62
Gene Name
cancer/testis antigen 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102658.1 1079 Silent Mutation TCA,TCG S102S NP_001096128.1
XM_006720429.3 1079 Silent Mutation TCA,TCG S102S XP_006720492.1
Gene
THSD4
Gene Name
thrombospondin type 1 domain containing 4
There are no transcripts associated with this gene.

View Full Product Details