Product Details

SNP ID
rs144384099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:93205620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGGCATCAGAGCAATCCCGGA[A/G]GTGTGAGGTTGCAACAAATAGCTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105370980 PubMed Links

Gene Details

Gene
LOC105370980
Gene Name
putative uncharacterized protein UNQ9370/PRO34162
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011522308.2 Intron XP_011520610.1

View Full Product Details