Product Details

SNP ID
rs144404388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:70891973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCCTGCTTGGTCTCCCTCCTC[A/T]CCCCTCTTAGGTGCCGGGGCGGGCA
Phenotype
MIM: 612538
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRC49 PubMed Links

Gene Details

Gene
LRRC49
Gene Name
leucine rich repeat containing 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199017.2 461 Intron NP_001185946.1
NM_001199018.2 461 Intron NP_001185947.1
NM_001284357.1 461 Intron NP_001271286.1
NM_017691.4 461 Intron NP_060161.2
XM_005254492.4 461 Intron XP_005254549.1
XM_011521715.2 461 Intron XP_011520017.1
XM_011521717.2 461 Intron XP_011520019.1
XM_011521718.2 461 Intron XP_011520020.1
XM_017022357.1 461 Intron XP_016877846.1
XM_017022358.1 461 Intron XP_016877847.1
XM_017022359.1 461 Intron XP_016877848.1
Gene
THAP10
Gene Name
THAP domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020147.3 461 Silent Mutation GGA,GGT G100G NP_064532.1

View Full Product Details