Product Details
- SNP ID
-
rs144574012
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:78173663 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTCTGGATGTGGTAGGGCCGGGC[C/T]GCCGCGTTCATGTTGACCCTCCGGA
- Phenotype
-
MIM: 614362
MIM: 601149
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACSBG1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11072735] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACSBG1
- Gene Name
- acyl-CoA synthetase bubblegum family member 1
- Gene
- IDH3A
- Gene Name
- isocitrate dehydrogenase 3 (NAD(+)) alpha
There are no transcripts associated with this gene.
View Full Product Details