Product Details

SNP ID
rs144739548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45117580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCAAAAGATGGAAGAAGGCCC[A/G]GGCATCACGCCTGTAATCCCAGCAC
Phenotype
MIM: 606759 MIM: 612771 MIM: 612772
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUOX2 PubMed Links

Gene Details

Gene
DUOX2
Gene Name
dual oxidase 2
There are no transcripts associated with this gene.

Gene
DUOXA1
Gene Name
dual oxidase maturation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276264.1 3006 Missense Mutation CGG,TGG R463W NP_001263193.1
NM_001276265.1 3006 Missense Mutation CGG,TGG R418W NP_001263194.1
NM_001276266.1 3006 Intron NP_001263195.1
NM_001276267.1 3006 UTR 3 NP_001263196.1
NM_001276268.1 3006 UTR 3 NP_001263197.1
NM_144565.3 3006 Missense Mutation CGG,TGG R463W NP_653166.2
XM_006720743.3 3006 UTR 3 XP_006720806.1
XM_006720745.1 3006 UTR 3 XP_006720808.1
XM_006720746.1 3006 UTR 3 XP_006720809.1
XM_006720747.3 3006 UTR 3 XP_006720810.1
XM_006720749.1 3006 UTR 3 XP_006720812.1
XM_006720751.3 3006 UTR 3 XP_006720814.1
XM_006720752.3 3006 UTR 3 XP_006720815.2
XM_006720754.1 3006 UTR 3 XP_006720817.1
XM_006720755.1 3006 UTR 3 XP_006720818.1
XM_011522181.2 3006 UTR 3 XP_011520483.1
XM_011522182.1 3006 UTR 3 XP_011520484.1
XM_011522183.2 3006 UTR 3 XP_011520485.1
XM_011522184.1 3006 Intron XP_011520486.1
XM_011522185.1 3006 Intron XP_011520487.1
XM_011522186.2 3006 Intron XP_011520488.1
Gene
DUOXA2
Gene Name
dual oxidase maturation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207581.3 3006 Intron NP_997464.2
XM_017022180.1 3006 Intron XP_016877669.1

View Full Product Details