Product Details

SNP ID
rs144826830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:58599707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCAGAGCAATTCCCATAAGTAA[C/T]ACTGCCCACCAATGAGCCTAGAAAT
Phenotype
MIM: 602192
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM10 PubMed Links

Gene Details

Gene
ADAM10
Gene Name
ADAM metallopeptidase domain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110.3 2487 Silent Mutation GTA,GTG V681V NP_001101.1
NM_001320570.1 2487 Silent Mutation GTA,GTG V650V NP_001307499.1
XM_017021861.1 2487 Silent Mutation GTA,GTG V607V XP_016877350.1

View Full Product Details