Product Details

SNP ID
rs144929846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65196812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCTGGCGCTGGCCACCCCTGCT[C/T]GCTCGCTGCTGCCTCCTCGAGGGCA
Phenotype
MIM: 603489
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CILP PubMed Links

Gene Details

Gene
CILP
Gene Name
cartilage intermediate layer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003613.3 3655 Silent Mutation GCA,GCG A1158A NP_003604.3
XM_017022678.1 3655 Silent Mutation GCA,GCG A1185A XP_016878167.1
XM_017022679.1 3655 Silent Mutation GCA,GCG A1134A XP_016878168.1

View Full Product Details