Product Details

SNP ID
rs145067457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48197930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTTAGTTAAGGGTAACAGTCA[A/G]GTAATGTAGAGGGCAGGCACAGAGG
Phenotype
MIM: 600839
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTXN2 PubMed Links

Gene Details

Gene
CTXN2
Gene Name
cortexin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145668.1 Intron NP_001139140.1
XM_005254378.3 Intron XP_005254435.1
XM_006720504.2 Intron XP_006720567.1
XM_011521553.1 Intron XP_011519855.1
XM_017022178.1 Intron XP_016877667.1
Gene
SLC12A1
Gene Name
solute carrier family 12 member 1
There are no transcripts associated with this gene.

View Full Product Details