Product Details

SNP ID
rs145216577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42945332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTGAGGCAGAGTTGGAGCTCA[C/T]AGTAACTGCCAGTTGAATCCAAATA
Phenotype
MIM: 605981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBR1 PubMed Links

Gene Details

Gene
UBR1
Gene Name
ubiquitin protein ligase E3 component n-recognin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174916.2 5313 Silent Mutation CTA,CTG L1749L NP_777576.1

View Full Product Details