Product Details

SNP ID
rs145704934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101177595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTCCTACTTCCTGGCTCCTAAA[C/T]GTCTTCAAACCTGCCTGGACAACCT
Phenotype
MIM: 608183
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHSY1 PubMed Links

Gene Details

Gene
CHSY1
Gene Name
chondroitin sulfate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014918.4 1960 Silent Mutation ACA,ACG T734T NP_055733.2
XM_006720435.3 1960 Silent Mutation ACA,ACG T462T XP_006720498.1
XM_011521364.2 1960 Silent Mutation ACA,ACG T762T XP_011519666.1
XM_017022011.1 1960 Silent Mutation ACA,ACG T462T XP_016877500.1

View Full Product Details