Product Details

SNP ID
rs145837770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101177757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGAAGCCAGTTTTCTGAGTAAA[C/G]GCAAAATGGTTGTCACTGGGAACTT
Phenotype
MIM: 608183
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHSY1 PubMed Links

Gene Details

Gene
CHSY1
Gene Name
chondroitin sulfate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014918.4 1798 Silent Mutation GCC,GCG A680A NP_055733.2
XM_006720435.3 1798 Silent Mutation GCC,GCG A408A XP_006720498.1
XM_011521364.2 1798 Silent Mutation GCC,GCG A708A XP_011519666.1
XM_017022011.1 1798 Silent Mutation GCC,GCG A408A XP_016877500.1

View Full Product Details