Product Details
- SNP ID
-
rs145915972
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:39801246 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGCTCTTCTGGGGTGTTGCCCAA[C/G]GGGGGAAGCAGCCGCTTCTTGCTGT
- Phenotype
-
MIM: 612183
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPR176
PubMed Links
Gene Details
- Gene
- GPR176
- Gene Name
- G protein-coupled receptor 176
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001271854.1 |
2623 |
Silent Mutation |
CCC,CCG |
P477P |
NP_001258783.1 |
NM_001271855.1 |
2623 |
Silent Mutation |
CCC,CCG |
P433P |
NP_001258784.1 |
NM_007223.2 |
2623 |
Silent Mutation |
CCC,CCG |
P478P |
NP_009154.1 |
XM_011521167.2 |
2623 |
Silent Mutation |
CCC,CCG |
P446P |
XP_011519469.1 |
XM_011521168.2 |
2623 |
Silent Mutation |
CCC,CCG |
P446P |
XP_011519470.1 |
XM_017021873.1 |
2623 |
Silent Mutation |
CCC,CCG |
P446P |
XP_016877362.1 |
XM_017021874.1 |
2623 |
Silent Mutation |
CCC,CCG |
P315P |
XP_016877363.1 |
XM_017021875.1 |
2623 |
Silent Mutation |
CCC,CCG |
P315P |
XP_016877364.1 |
XM_017021876.1 |
2623 |
Silent Mutation |
CCC,CCG |
P315P |
XP_016877365.1 |
XM_017021877.1 |
2623 |
Silent Mutation |
CCC,CCG |
P315P |
XP_016877366.1 |
XM_017021878.1 |
2623 |
Silent Mutation |
CCC,CCG |
P315P |
XP_016877367.1 |
View Full Product Details