Product Details
- SNP ID
-
rs146049960
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:42726314 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGCAGATGCCACAGCTCACCTGG[C/T]GGCACCGCAGCGTCTGGCCCAGCTG
- Phenotype
-
MIM: 607465
MIM: 614642
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CDAN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28661826] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDAN1
- Gene Name
- codanin 1
- Gene
- STARD9
- Gene Name
- StAR related lipid transfer domain containing 9
There are no transcripts associated with this gene.
View Full Product Details