Product Details

SNP ID
rs146217804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50182615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTGCGGGCGTTCCTGGAGAAAG[C/T]GCGCCAGACGCCACACAAGCCTTTT
Phenotype
MIM: 609123 MIM: 603247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 420 Intron NP_079113.2
XM_011522046.2 420 Intron XP_011520348.1
XM_011522047.2 420 Intron XP_011520349.1
XM_011522048.1 420 Intron XP_011520350.1
XM_011522049.2 420 Intron XP_011520351.1
XM_011522051.2 420 Intron XP_011520353.1
XM_011522052.2 420 Intron XP_011520354.1
XM_011522053.1 420 Intron XP_011520355.1
XM_011522056.2 420 Intron XP_011520358.2
XM_011522058.2 420 Intron XP_011520360.1
XM_011522059.1 420 Intron XP_011520361.1
XM_011522060.1 420 Intron XP_011520362.1
XM_011522061.1 420 Intron XP_011520363.1
XM_011522062.1 420 Intron XP_011520364.1
XM_011522063.1 420 Intron XP_011520365.1
XM_011522064.1 420 Intron XP_011520366.1
XM_011522069.2 420 Intron XP_011520371.1
XM_011522070.1 420 Intron XP_011520372.1
XM_017022587.1 420 Intron XP_016878076.1
XM_017022588.1 420 Intron XP_016878077.1
XM_017022589.1 420 Intron XP_016878078.1
XM_017022590.1 420 Intron XP_016878079.1
XM_017022591.1 420 Intron XP_016878080.1
XM_017022592.1 420 Intron XP_016878081.1
XM_017022593.1 420 Intron XP_016878082.1
XM_017022594.1 420 Intron XP_016878083.1
XM_017022595.1 420 Intron XP_016878084.1
XM_017022596.1 420 Intron XP_016878085.1
XM_017022597.1 420 Intron XP_016878086.1
XM_017022598.1 420 Intron XP_016878087.1
XM_017022599.1 420 Intron XP_016878088.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159629.1 420 Missense Mutation GCG,GTG A63V NP_001153101.1
NM_003645.3 420 Missense Mutation GCG,GTG A63V NP_003636.2

View Full Product Details